Merck
  • 2-[(Carboxymethyl)sulfanyl]-4-oxo-4-arylbutanoic acids selectively suppressed proliferation of neoplastic human HeLa cells. A SAR/QSAR study.
2-[(Carboxymethyl)sulfanyl]-4-oxo-4-arylbutanoic acids selectively suppressed proliferation of neoplastic human HeLa cells. A SAR/QSAR study.
Journal of medicinal chemistry (2005-08-19)
Branko J Drakulić, Zorica D Juranić, Tatjana P Stanojković, Ivan O Juranić
ABSTRACT
A series of twenty alkyl-, halo-, and methoxy-aryl-substituted 2-[(carboxymethyl)sulfanyl]-4-oxo-4-arylbutanoic acids were synthesized. The new compounds, called CSAB, suppressed proliferation of human cervix carcinoma, HeLa cells, in vitro in a concentration range of 0.644 to 29.48 microM/L. Two compounds exhibit antiproliferative activity in sub-micromolar concentrations. Five compounds act in low micromolar concentrations (<2 microM/L). The most active compounds exert lower cytotoxicity toward healthy human peripheral blood mononuclear cells (PBMC and PBMC+PHA) (selectivity indexes > 10). A strong structure-activity relationship, using estimated log P values and BCUT descriptors, was observed.
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Thioglycolic acid, ≥99%