Merck
  • Synthesis and in vitro antibiotic activity of 16-membered 9-O-arylalkyloxime macrolides.
Synthesis and in vitro antibiotic activity of 16-membered 9-O-arylalkyloxime macrolides.
Bioorganic & medicinal chemistry letters (2005-12-14)
Hong Fu, Saul Marquez, Xiangrong Gu, Leonard Katz, David C Myles
ABSTRACT
A series of novel 9-O-arylalkyloxime analogs based on three different 16-membered macrolide scaffolds-5-O-mycaminosyltylonolide (OMT), tilmicosin, and 20-deoxy-20-(3,5-dimethyl-1-piperidin-1-yl)-OMT-was synthesized. In vitro antibiotic activities were assayed against Gram-positive Streptococcus pneumoniae and Staphylococcus aureus and Gram-negative Haemophilus influenzae bacterial strains. Analogs derived from OMT (3-15) showed similar or better antibacterial activities against macrolide-susceptible strains and enhanced activities against macrolide-resistant strains compared with erythromycin A, tylosin, or OMT. Similar results were observed for tilmicosin 9-O-arylalkyloxime analogs (18-24). In contrast, most of the 20-deoxy-20-(3,5-dimethyl-1-piperidin-1-yl)-OMT analogs (25-33) showed reduced antibacterial activities compared with OMT. Ribosome-binding studies were performed on compounds 12 (OMT derivative), 20 (tilmicosin derivative), and 29 [20-deoxy-20-(3,5-dimethyl-1-piperidin-1-yl)-OMT derivative]. It was found that these compounds interacted with both domain V and domain II of the Escherichia coli 23S rRNA.
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications