Merck
  • Tactile modulation of memory and anxiety requires dentate granule cells along the dorsoventral axis.
Tactile modulation of memory and anxiety requires dentate granule cells along the dorsoventral axis.
Nature communications (2020-11-29)
Chi Wang, Hui Liu, Kun Li, Zhen-Zhen Wu, Chen Wu, Jing-Ying Yu, Qian Gong, Ping Fang, Xing-Xing Wang, Shu-Min Duan, Hao Wang, Yan Gu, Ji Hu, Bing-Xing Pan, Mathias V Schmidt, Yi-Jun Liu, Xiao-Dong Wang
ABSTRACT
Touch can positively influence cognition and emotion, but the underlying mechanisms remain unclear. Here, we report that tactile experience enrichment improves memory and alleviates anxiety by remodeling neurons along the dorsoventral axis of the dentate gyrus (DG) in adult mice. Tactile enrichment induces differential activation and structural modification of neurons in the dorsal and ventral DG, and increases the presynaptic input from the lateral entorhinal cortex (LEC), which is reciprocally connected with the primary somatosensory cortex (S1), to tactile experience-activated DG neurons. Chemogenetic activation of tactile experience-tagged dorsal and ventral DG neurons enhances memory and reduces anxiety respectively, whereas inactivation of these neurons or S1-innervated LEC neurons abolishes the beneficial effects of tactile enrichment. Moreover, adulthood tactile enrichment attenuates early-life stress-induced memory deficits and anxiety-related behavior. Our findings demonstrate that enriched tactile experience retunes the pathway from S1 to DG and enhances DG neuronal plasticity to modulate cognition and emotion.
Sigma-Aldrich
SIGMAFAST DAB with Metal Enhancer, tablet
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-496 mreelin, clone G10, clone G10, Chemicon®, from mouse