Merck
  • Correlation between olfactory receptor cell type and function in the channel catfish.
Correlation between olfactory receptor cell type and function in the channel catfish.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2003-10-17)
Anne Hansen, Shane H Rolen, Karl Anderson, Yasuhiro Morita, John Caprio, Thomas E Finger
ABSTRACT
The olfactory epithelium of fish contains three intermingled types of olfactory receptor neurons (ORNs): ciliated, microvillous, and crypt. The present experiments were undertaken to test whether the different types of ORNs respond to different classes of odorants via different families of receptor molecules and G-proteins corresponding to the morphology of the ORN. In catfish, ciliated ORNs express OR-type receptors and Galpha(olf). Microvillous ORNs are heterogeneous, with many expressing Galpha(q)/11, whereas crypt ORNs express Galpha(o). Retrograde tracing experiments show that ciliated ORNs project predominantly to regions of the olfactory bulb (OB) that respond to bile salts (medial) and amino acids (ventral) (Nikonov and Caprio, 2001). In contrast, microvillous ORNs project almost entirely to the dorsal surface of the OB, where responses to nucleotides (posterior OB) and amino acids (anterior OB) predominate. These anatomical findings are consistent with our pharmacological results showing that forskolin (which interferes with Galpha(olf)/cAMP signaling) blocks responses to bile salts and markedly reduces responses to amino acids. Conversely, U-73122 and U-73343 (which interfere with Galpha(q)/11/phospholipase C signaling) diminish amino acid responses but leave bile salt and nucleotide responses essentially unchanged. In summary, our results indicate that bile salt odorants are detected predominantly by ciliated ORNs relying on the Galpha(olf)/cAMP transduction cascade. Nucleotides are detected by microvillous ORNs using neither Galpha(olf)/cAMP nor Galpha(q)/11/PLC cascades. Finally, amino acid odorants activate both ciliated and microvillous ORNs but via different transduction pathways in the two types of cells.
Sigma-Aldrich
Anti-Gqα-and G11α-Subunits, C-Terminal Rabbit pAb, liquid, Calbiochem®