Merck
  • Directed evolution of APEX2 for electron microscopy and proximity labeling.
Directed evolution of APEX2 for electron microscopy and proximity labeling.
Nature methods (2014-11-25)
Stephanie S Lam, Jeffrey D Martell, Kimberli J Kamer, Thomas J Deerinck, Mark H Ellisman, Vamsi K Mootha, Alice Y Ting
ABSTRACT
APEX is an engineered peroxidase that functions as an electron microscopy tag and a promiscuous labeling enzyme for live-cell proteomics. Because limited sensitivity precludes applications requiring low APEX expression, we used yeast-display evolution to improve its catalytic efficiency. APEX2 is far more active in cells, enabling the use of electron microscopy to resolve the submitochondrial localization of calcium uptake regulatory protein MICU1. APEX2 also permits superior enrichment of endogenous mitochondrial and endoplasmic reticulum membrane proteins.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Anti-Calreticulin Mouse mAb (FMC75), liquid, clone FMC75, Calbiochem®
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution