Merck
  • Antigen-specific T-cell responses in patients with non-IgE-mediated gastrointestinal food allergy are predominantly skewed to T(H)2.
Antigen-specific T-cell responses in patients with non-IgE-mediated gastrointestinal food allergy are predominantly skewed to T(H)2.
The Journal of allergy and clinical immunology (2012-10-23)
Hideaki Morita, Ichiro Nomura, Kanami Orihara, Koichi Yoshida, Akira Akasawa, Hiroshi Tachimoto, Yoshikazu Ohtsuka, Yoshiyuki Namai, Masaki Futamura, Tetsuo Shoda, Akio Matsuda, Norio Kamemura, Hiroshi Kido, Takao Takahashi, Yukihiro Ohya, Hirohisa Saito, Kenji Matsumoto
Sigma-Aldrich
κ-Casein from bovine milk, ≥70% (PAGE), lyophilized powder