Merck
  • Gene Copy Number Estimation from Targeted Next-Generation Sequencing of Prostate Cancer Biopsies: Analytic Validation and Clinical Qualification.
Gene Copy Number Estimation from Targeted Next-Generation Sequencing of Prostate Cancer Biopsies: Analytic Validation and Clinical Qualification.
Clinical cancer research : an official journal of the American Association for Cancer Research (2017-07-29)
George Seed, Wei Yuan, Joaquin Mateo, Suzanne Carreira, Claudia Bertan, Maryou Lambros, Gunther Boysen, Roberta Ferraldeschi, Susana Miranda, Ines Figueiredo, Ruth Riisnaes, Mateus Crespo, Daniel Nava Rodrigues, Eric Talevich, Dan R Robinson, Lakshmi P Kunju, Yi-Mi Wu, Robert Lonigro, Shahneen Sandhu, Arul M Chinnaiyan, Johann S de Bono
ABSTRACT
Purpose: Precise detection of copy number aberrations (CNA) from tumor biopsies is critically important to the treatment of metastatic prostate cancer. The use of targeted panel next-generation sequencing (NGS) is inexpensive, high throughput, and easily feasible, allowing single-nucleotide variant calls, but CNA estimation from this remains challenging.Experimental Design: We evaluated CNVkit for CNA identification from amplicon-based targeted NGS in a cohort of 110 fresh castration-resistant prostate cancer biopsies and used capture-based whole-exome sequencing (WES), array comparative genomic hybridization (aCGH), and FISH to explore the viability of this approach.Results: We showed that this method produced highly reproducible CNA results (r = 0.92), with the use of pooled germline DNA as a coverage reference supporting precise CNA estimation. CNA estimates from targeted NGS were comparable with WES (r = 0.86) and aCGH (r = 0.7); for key selected genes (BRCA2, MYC, PIK3CA, PTEN, and RB1), CNA estimation correlated well with WES (r = 0.91) and aCGH (r = 0.84) results. The frequency of CNAs in our population was comparable with that previously described (i.e., deep deletions: BRCA2 4.5%; RB1 8.2%; PTEN 15.5%; amplification: AR 45.5%; gain: MYC 31.8%). We also showed, utilizing FISH, that CNA estimation can be impacted by intratumor heterogeneity and demonstrated that tumor microdissection allows NGS to provide more precise CNA estimates.Conclusions: Targeted NGS and CNVkit-based analyses provide a robust, precise, high-throughput, and cost-effective method for CNA estimation for the delivery of more precise patient care. Clin Cancer Res; 23(20); 6070-7. ©2017 AACR.
Sigma-Aldrich
GenomePlex® Complete Whole Genome Amplification (WGA) Kit, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue