Merck
CDK2 regulates the NRF1/Ehmt1 axis during meiotic prophase I.
The Journal of cell biology (2019-07-28)
Nathan Palmer, S Zakiah A Talib, Chandrahas Koumar Ratnacaram, Diana Low, Xavier Bisteau, Joanna Hui Si Lee, Elisabeth Pfeiffenberger, Heike Wollmann, Joel Heng Loong Tan, Sheena Wee, Radoslaw Sobota, Jayantha Gunaratne, Daniel M Messerschmidt, Ernesto Guccione, Philipp Kaldis
ABSTRACT
Meiosis generates four genetically distinct haploid gametes over the course of two reductional cell divisions. Meiotic divisions are characterized by the coordinated deposition and removal of various epigenetic marks. Here we propose that nuclear respiratory factor 1 (NRF1) regulates transcription of euchromatic histone methyltransferase 1 (EHMT1) to ensure normal patterns of H3K9 methylation during meiotic prophase I. We demonstrate that cyclin-dependent kinase (CDK2) can bind to the promoters of a number of genes in male germ cells including that of Ehmt1 through interaction with the NRF1 transcription factor. Our data indicate that CDK2-mediated phosphorylation of NRF1 can occur at two distinct serine residues and negatively regulates NRF1 DNA binding activity in vitro. Furthermore, induced deletion of Cdk2 in spermatocytes results in increased expression of many NRF1 target genes including Ehmt1 We hypothesize that the regulation of NRF1 transcriptional activity by CDK2 may allow the modulation of Ehmt1 expression, therefore controlling the dynamic methylation of H3K9 during meiotic prophase.
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Roche
DIG Oligonucleotide 3′-End Labeling Kit, 2nd generation, sufficient for 25 labeling reactions (100 pmol of oligonucleotides per assay; 1 ug of a 30-mer oligonucleotide), storage condition avoid repeated freeze/thaw cycles
Sigma-Aldrich
UNC0642, ≥98% (HPLC)
Sigma-Aldrich
ApopTag Plus Peroxidase In Situ Apoptosis Kit, The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling & detecting DNA strand breaks by the indirect TUNEL method.
Sigma-Aldrich
Tamoxifen, ≥99%