Merck
  • Effects of parental exposure to glyphosate-based herbicides on embryonic development and oxidative status: a long-term experiment in a bird model.
Effects of parental exposure to glyphosate-based herbicides on embryonic development and oxidative status: a long-term experiment in a bird model.
Scientific reports (2020-04-15)
Suvi Ruuskanen, Miia J Rainio, Maiju Uusitalo, Kari Saikkonen, Marjo Helander
ABSTRACT
Controversial glyphosate-based herbicides (GBHs) are the most frequently used herbicides globally. GBH residues are detected in soil, water, crops, and food products, potentially exposing non-target organisms to health risks; these organisms include wildlife, livestock, and humans. However, the potential for GBH-related parental effects are poorly understood. In the case of birds, GBHs may be transferred directly from mothers to eggs, or they may indirectly influence offspring performance by altered maternal resource allocation to eggs. We experimentally exposed a parental generation of Japanese quails (Coturnix japonica) to GBHs (200 mg/kg feed) or respective controls. Glyphosate residues were found in eggs (ca 0.76 kg/mg). Embryonic development tended to be poorer in the eggs of GBH-exposed parents (76% of eggs showed normal development) compared to control parents (89% normal eggs). Embryonic brain tissue from GBH-exposed parents tended to express more lipid damage (20% higher), yet other biomarkers showed no apparent differences. We detected no differences in egg quality (egg, yolk, or shell mass, egg hormone concentration) across the treatment groups. Given this is the first long-term study testing parental effects of GBHs with birds, more studies are needed characterizing GBH-associated changes in maternal allocation and for example epigenetic programming.
Sigma-Aldrich
Glutathione-S-Transferase (GST) Assay Kit, sufficient for 500 multiwell tests
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests