Merck
  • USP52 regulates DNA end resection and chemosensitivity through removing inhibitory ubiquitination from CtIP.
USP52 regulates DNA end resection and chemosensitivity through removing inhibitory ubiquitination from CtIP.
Nature communications (2020-10-25)
Ming Gao, Guijie Guo, Jinzhou Huang, Jake A Kloeber, Fei Zhao, Min Deng, Xinyi Tu, Wootae Kim, Qin Zhou, Chao Zhang, Ping Yin, Kuntian Luo, Zhenkun Lou
ABSTRACT
Human C-terminal binding protein (CtBP)-interacting protein (CtIP) is a central regulator to initiate DNA end resection and homologous recombination (HR). Several studies have shown that post-translational modifications control the activity or expression of CtIP. However, it remains unclear whether and how cells restrain CtIP activity in unstressed cells and activate CtIP when needed. Here, we identify that USP52 directly interacts with and deubiquitinates CtIP, thereby promoting DNA end resection and HR. Mechanistically, USP52 removes the ubiquitination of CtIP to facilitate the phosphorylation and activation of CtIP at Thr-847. In addition, USP52 is phosphorylated by ATM at Ser-1003 after DNA damage, which enhances the catalytic activity of USP52. Furthermore, depletion of USP52 sensitizes cells to PARP inhibition in a CtIP-dependent manner in vitro and in vivo. Collectively, our findings reveal the key role of USP52 and the regulatory complexity of CtIP deubiquitination in DNA repair.
Sigma-Aldrich
MISSION® esiRNA, targeting human RBBP8
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)