Merck
  • Synthesis and enzymatic evaluation of novel partially fluorinated thiol dual ACE/NEP inhibitors.
Synthesis and enzymatic evaluation of novel partially fluorinated thiol dual ACE/NEP inhibitors.
Bioorganic & medicinal chemistry letters (2009-07-15)
Francesca Olimpieri, Simone Tambaro, Santos Fustero, Paolo Lazzari, Maria Sanchez-Roselló, Luca Pani, Alessandro Volonterio, Matteo Zanda
ABSTRACT
A novel family of peptidomimetics incorporating fluoroalkyl groups, mainly a trifluoromethyl, in alpha-position to a zinc(II)-binding thiol function, was synthesized in solution as well as in solid-phase. These compounds showed inhibitory potency in the nanomolar range against both angiotensin-converting enzyme (ACE) and neutral endopeptidase (NEP), whereas no inhibition of endothelin-converting enzyme-1 (ECE-1) was observed. The trifluoromethyl-derivatives were more potent than the parent unfluorinated analogues in the case of ACE, and less potent in the case of NEP.
Sigma-Aldrich
DL-Thiorphan, ≥98% (TLC)
Sigma-Aldrich
Captopril, meets USP testing specifications
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder