Merck
  • Activity-based protein profiling of rice (Oryza sativa L.) bran serine hydrolases.
Activity-based protein profiling of rice (Oryza sativa L.) bran serine hydrolases.
Scientific reports (2020-09-18)
Achintya Kumar Dolui, Arun Kumar Vijayakumar, Ram Rajasekharan, Panneerselvam Vijayaraj
ABSTRACT
Rice bran is an underutilized agricultural by-product with economic importance. The unique phytochemicals and fatty acid compositions of bran have been targeted for nutraceutical development. The endogenous lipases and hydrolases are responsible for the rapid deterioration of rice bran. Hence, we attempted to provide the first comprehensive profiling of active serine hydrolases (SHs) present in rice bran proteome by activity-based protein profiling (ABPP) strategy. The active site-directed fluorophosphonate probe (rhodamine and biotin-conjugated) was used for the detection and identification of active SHs. ABPP revealed 55 uncharacterized active-SHs and are representing five different known enzyme families. Based on motif and domain analyses, one of the uncharacterized and miss annotated SHs (Os12Ssp, storage protein) was selected for biochemical characterization by overexpressing in yeast. The purified recombinant protein authenticated the serine protease activity in time and protein-dependent studies. Os12Ssp exhibited the maximum activity at a pH between 7.0 and 8.0. The protease activity was inhibited by the covalent serine protease inhibitor, which suggests that the ABPP approach is indeed reliable than the sequence-based annotations. Collectively, the comprehensive knowledge generated from this study would be useful in expanding the current understanding of rice bran SHs and paves the way for better utilization/stabilization of rice bran.
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Trypsin from porcine pancreas, Proteomics Grade, BioReagent, Dimethylated