Merck
  • Improvement of resistant starch content and baking quality of cross-linked soft rice flour.
Improvement of resistant starch content and baking quality of cross-linked soft rice flour.
Food science and biotechnology (2020-12-08)
Chae Eun Lee, Junhee No, Kyongae Lee, Malshick Shin
ABSTRACT
To increase resistant starch (RS) content of rice flour, soft and normal rice flours were cross-linked with different flour concentrations (40, 50%). RS contents, morphology, and baking qualities of cross-linked Singil rice flour (CSRF) and Hopyeong rice flour (CHRF) were compared. Amylose and protein contents of Singil flour were higher than those of Hopyeong flour. The protein content of CSRF maintained but that of CHRF reduced. Although the RS content increased after cross-linking, the degree of RS was higher 50% flour than 40% flour. The shape of cross-linked rice flour particles changed into starch granules with debris. The overall quality and texture of CSRF40 cupcake showed the highest scores by preference test. These results suggest that CSRF can be used as a high RS rice flour substitute for wheat flour, because soft rice is easy to make flour.
Sigma-Aldrich
Protease from Bacillus licheniformis, lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Supelco
Amyloglucosidase solution from Aspergillus niger, for use in Total Dietary Fiber Assay, TDF-100A