Merck
  • Proteolysis of whey protein isolates in nanoemulsion systems: Impact of nanoemulsification and additional synthetic emulsifiers.
Proteolysis of whey protein isolates in nanoemulsion systems: Impact of nanoemulsification and additional synthetic emulsifiers.
Food chemistry (2021-03-02)
Patrycja Szumała, Aneta Pacyna-Kuchta, Andrzej Wasik
ABSTRACT
Nanoemulsions are currently of interest in the functional food sector because their small droplet size (100-500 nm) provides a number of potential advantages over conventional emulsions. This study concerned the behavior of nanoemulsions stabilized with whey proteins and two synthetic emulsifiers (Tween 80 and Croduret), and exposed to conditions simulating the human upper gastrointestinal tract. In particular, the effect of synthetic emulsifiers (food additives) on the interfacial composition and digestion rate of milk proteins at the interface of nanoemulsions was determined. The results indicate that the protein was partially co-absorbed with only one synthetic emulsifier (Croduret) at the interface, which made protein more resistant to digestion in the nanoemulsion system. This suggests that the degree of protein digestion can be controlled by appropriate selection of synthetic emulsifiers and presenting the protein in nanoemulsion system.
Product Description
Sodium taurocholate, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Sodium glycodeoxycholate, BioXtra, ≥97% (HPLC)
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein