Merck
FTO regulates ocular angiogenesis via m6A-YTHDF2-dependent mechanism.
Experimental eye research (2020-06-13)
Kun Shan, Rong-Mei Zhou, Jun Xiang, Ya-Nan Sun, Chang Liu, Meng-Wei Lv, Jian-Jiang Xu
ABSTRACT
Pathological ocular angiogenesis commonly results in visual impairment or even blindness. Unveiling the mechanisms of pathological angiogenesis is critical to identify the regulators and develop effective targeted therapies. Here, we used corneal neovascularization (CNV) model to investigate the mechanism of pathological ocular angiogenesis. We show that N6-methyladenosine (m6A) mRNA demethylation mediated by fat mass- and obesity-associated protein (FTO) could regulate endothelial cell (EC) function and pathological angiogenesis during CNV. FTO levels are increased in neovascularized corneas and ECs under pathological conditions. In vitro silencing of FTO in ECs results in reduced cellular proliferation, migration, and tube formation under both basal and pathological conditions. Furthermore, FTO silencing attenuates suture-induced CNV in vivo. Mechanically, FTO silencing in ECs could increase m6A methylation levels in critical pro-angiogenic genes, such as FAK, leading to decreased RNA stability and increased RNA decay through m6A reader YTHDF2. Our study demonstrates that FTO regulates pathological ocular angiogenesis by controlling EC function in an m6A-YTHDF2-dependent manner.
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Triton X-100, laboratory grade
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
Methylene blue, certified by the BSC