Merck
  • Hopane-type triterpenes and binaphthopyrones from the scale insect pathogenic fungus Aschersonia paraphysata BCC 11964.
Hopane-type triterpenes and binaphthopyrones from the scale insect pathogenic fungus Aschersonia paraphysata BCC 11964.
Journal of natural products (2010-04-07)
Masahiko Isaka, Arunrat Yangchum, Pranee Rachtawee, Somjit Komwijit, Apisara Lutthisungneon
ABSTRACT
Two new triterpenes, 17(21)-hopene-6alpha,12beta-diol (1) and 17(21)-hopen-12beta-ol (2), the known 17(21)-hopen-6alpha-ol (zeorinin, 3), and two new biarylic dihydronaphthopyrones, aschernaphthopyrones A (4) and B (5), were isolated from the scale insect pathogenic fungus Aschersonia paraphysata BCC 11964. Hopene 1 and aschernaphthopyrone A (4) exhibited antimalarial activity with IC(50) values of 15 and 7.3 microM, respectively.
Product Description
Supelco
Hop-22(29)-ene solution, 0.1 mg/mL in isooctane, analytical standard
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)