Merck
  • Evaluation of ionic equilibria in mixed-buffer isothermal titration calorimetry and continuously stirred tank reactors.
Evaluation of ionic equilibria in mixed-buffer isothermal titration calorimetry and continuously stirred tank reactors.
International journal of pharmaceutics (2020-12-15)
Harrison B Rose, Madison M Wilber, Andreas S Bommarius
ABSTRACT
Determination of an equilibrium pH value in complex aqueous solution and deconvolution of this equilibrium to evaluate phenomena related to mixing, dilution, or progress of reaction is increasingly important in areas ranging from water quality to pharmaceutical formulations and manufacturing. Linearization of pH problems by simple algebraic substitution enables equilibria within complex buffered aqueous solutions to be modeled as an eigenvalue problem. This formulation approach makes rigorous determination of equilibrium pH values and reactor dynamics more accessible than with previous calculation methods, even when activity coefficients and non-ideality are considered. This work demonstrates how such calculations can enable detailed modeling of enthalpic changes in an isothermal titration calorimeter. In support of this work, the acid dissociation constants for three furancarboxylic acids (2-furancarboxylic acid, FA; 5-formyl-2-furancarboxylic acid, FFA; and 2,5-furandicarboxylic acid, FDCA), two of them novel, were determined and compared with multi-wavelength ultraviolet-visible spectrophotometry. The thermodynamic pKa values were determined to be 3.1 for FA, 2.2 for FFA, and 2.1 and 3.4 for the first and second ionization steps of FDCA, respectively.
Sigma-Aldrich
2,5-Furandicarboxylic acid, 97%