Merck
  • Determination of Oxygen Respiration Rates in Wetted Developmentally Arrested Spores of Streptomyces Species
Determination of Oxygen Respiration Rates in Wetted Developmentally Arrested Spores of Streptomyces Species
Bio-protocol (4)
Fischer M, et al.