Merck
  • Tissue Blot Immunoassay for Detection of Tomato spotted wilt virus in Ranunculus asiaticus and Other Ornamentals.
Tissue Blot Immunoassay for Detection of Tomato spotted wilt virus in Ranunculus asiaticus and Other Ornamentals.
Plant disease (2003-06-01)
A E Whitfield, L R Campbell, J L Sherwood, D E Ullman
ABSTRACT
A tissue blot immunoassay (TBIA) was developed to detect Tomato spotted wilt virus (TSWV) in Ranunculus asiaticus tubers and other ornamentals. TBIA was comparable to double-antibody sandwich enzyme-linked immunosorbent assay for accuracy and reliability. A nondestructive sampling method was used with R. asiaticus tubers to determine: (i) the relationship between tuber infection and size; (ii) the distribution of TSWV in tubers; and (iii) the relationship between tuber infection and tuber germination. Small tubers had a higher percentage (44%) of infection than large tubers (19%). When destructive sampling was acceptable, the central stem tissue was the most reliable to test. TSWV infection was associated with a significant reduction of tuber germination. Among the tubers that tested positive for TSWV infection, 48% of those that germinated produced foliage in which TSWV was detected. The remaining 52% of the infected tubers planted that germinated developed into plants that were asymptomatic and in which TSWV was not detectable after germination. Only 4% of tubers that tested negative produced infected plants after germination. Our results indicate that TBIA can be used in TSWV management programs to identify infected plants and to index tuber crops.
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet