Merck
  • A Method to Detect the Binding of Hyper-Glycosylated Fragment Crystallizable (Fc) Region of Human IgG1 to Glycan Receptors.
A Method to Detect the Binding of Hyper-Glycosylated Fragment Crystallizable (Fc) Region of Human IgG1 to Glycan Receptors.
Methods in molecular biology (Clifton, N.J.) (2018-12-13)
Patricia Blundell, Richard Pleass
ABSTRACT
Engineering the fragment crystallizable (Fc) of human IgG can bring improved effector functions to monoclonal antibodies and Fc-fusion-based medicines and vaccines. Such Fc-effector functions are largely controlled by posttranslational modifications (PTMs) within the Fc, including the addition of glycans that introduce structural and functional heterogeneity to this class of therapeutic. Here, we describe a detailed method to allow the detection of hyper-sialylated Fcs to glycan receptors that will facilitate the future development of new mAbs and Fc-fragment therapies and vaccines.
Sigma-Aldrich
Carbonate-Bicarbonate Buffer, capsule
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
SIGMAFAST p-Nitrophenyl phosphate Tablets, tablet, To prepare 20 mL