  • Search Results
  • Redox feedback regulation of ANAC089 signaling alters seed germination and stress response.
Redox feedback regulation of ANAC089 signaling alters seed germination and stress response.
Cell reports (2021-06-17)
Pablo Albertos, Kiyoshi Tatematsu, Isabel Mateos, Inmaculada Sánchez-Vicente, Alejandro Fernández-Arbaizar, Kazumi Nakabayashi, Eiji Nambara, Marta Godoy, José M Franco, Roberto Solano, Davide Gerna, Thomas Roach, Wolfgang Stöggl, Ilse Kranner, Carlos Perea-Resa, Julio Salinas, Oscar Lorenzo
ABSTRACT
The interplay between the phytohormone abscisic acid (ABA) and the gasotransmitter nitric oxide (NO) regulates seed germination and post-germinative seedling growth. We show that GAP1 (germination in ABA and cPTIO 1) encodes the transcription factor ANAC089 with a critical membrane-bound domain and extranuclear localization. ANAC089 mutants lacking the membrane-tethered domain display insensitivity to ABA, salt, and osmotic and cold stresses, revealing a repressor function. Whole-genome transcriptional profiling and DNA-binding specificity reveals that ANAC089 regulates ABA- and redox-related genes. ANAC089 truncated mutants exhibit higher NO and lower ROS and ABA endogenous levels, alongside an altered thiol and disulfide homeostasis. Consistently, translocation of ANAC089 to the nucleus is directed by changes in cellular redox status after treatments with NO scavengers and redox-related compounds. Our results reveal ANAC089 to be a master regulator modulating redox homeostasis and NO levels, able to repress ABA synthesis and signaling during Arabidopsis seed germination and abiotic stress.
Sigma-Aldrich
CelLytic PN Isolation/Extraction Kit, For plant leaves
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Triton X-100, for molecular biology
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
D-Mannitol, ≥98%
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Anti-Actin (plant) antibody, Mouse monoclonal, clone 10-B3 (MAbGPa), purified from hybridoma cell culture