Merck
Gating movements and ion permeation in HCN4 pacemaker channels.
Molecular cell (2021-06-25)
Andrea Saponaro, Daniel Bauer, M Hunter Giese, Paolo Swuec, Alessandro Porro, Federica Gasparri, Atiyeh Sadat Sharifzadeh, Antonio Chaves-Sanjuan, Laura Alberio, Giacomo Parisi, Gabriele Cerutti, Oliver B Clarke, Kay Hamacher, Henry M Colecraft, Filippo Mancia, Wayne A Hendrickson, Steven A Siegelbaum, Dario DiFrancesco, Martino Bolognesi, Gerhard Thiel, Bina Santoro, Anna Moroni
ABSTRACT
The HCN1-4 channel family is responsible for the hyperpolarization-activated cation current If/Ih that controls automaticity in cardiac and neuronal pacemaker cells. We present cryoelectron microscopy (cryo-EM) structures of HCN4 in the presence or absence of bound cAMP, displaying the pore domain in closed and open conformations. Analysis of cAMP-bound and -unbound structures sheds light on how ligand-induced transitions in the channel cytosolic portion mediate the effect of cAMP on channel gating and highlights the regulatory role of a Mg2+ coordination site formed between the C-linker and the S4-S5 linker. Comparison of open/closed pore states shows that the cytosolic gate opens through concerted movements of the S5 and S6 transmembrane helices. Furthermore, in combination with molecular dynamics analyses, the open pore structures provide insights into the mechanisms of K+/Na+ permeation. Our results contribute mechanistic understanding on HCN channel gating, cyclic nucleotide-dependent modulation, and ion permeation.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein