Merck
  • Fluorescent tagging of endogenous proteins with CRISPR/Cas9 in primary mouse neural stem cells.
Fluorescent tagging of endogenous proteins with CRISPR/Cas9 in primary mouse neural stem cells.
STAR protocols (2021-08-26)
Christopher S Morrow, Tiaira J Porter, Darcie L Moore
ABSTRACT
Although exogenous overexpression of a protein fused to a fluorescent tag can provide insight for the protein's function, it also can produce artifacts attributed to its upregulation and may not fully report the endogenous regulation of the protein of interest. To circumvent these issues, we adapted a protocol to label endogenous proteins with fluorescent tags in primary adult mouse neural stem cells in vitro. Here, we describe reagent construction, reagent delivery, and a screening strategy to isolate edited cells. For complete details on the use and execution of this protocol, please refer to Morrow et al. (2020).
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture