Merck
  • Assays for studying normal versus suppressive ERAD-associated retrotranslocation pathways in yeast.
Assays for studying normal versus suppressive ERAD-associated retrotranslocation pathways in yeast.
STAR protocols (2021-07-20)
Satarupa Bhaduri, Sonya E Neal
ABSTRACT
In S. cerevisiae, we identified rhomboid pseudoprotease Dfm1 as the major mediator for removing or retrotranslocating misfolded membrane substrates from the ER (endoplasmic reticulum). Long-standing challenges with rapid suppression of dfm1-null cells have limited the biochemical study of Dfm1's role in ER protein quality control. Here, we provide a protocol for the generation and handling of dfm1-null cells and procedures for studying normal vs. suppressive alternative retrotranslocation pathways. Our methods can be utilized to study other components involved in retrotranslocation. For complete information on the generation and use of this protocol, please refer to Neal et al. (2017, 2018); Neal et al. (2019); Neal et al. (2020).
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Yeast Nitrogen Base Without Amino Acids, Yeast classification medium used for selecting yeasts based on amino acid and carbohydrate requirements
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
N-p-Tosyl-L-phenylalanine chloromethyl ketone, ≥97% (TLC), powder
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
MG-132, ≥95% by HPLC, Potent, reversible, and cell-permeable proteasome inhibitor (Ki = 4 nM).