Merck
  • Electrophoretic Mobility Shift Assay of in vitro Phosphorylated RNA Polymerase II Carboxyl-terminal Domain Substrates.
Electrophoretic Mobility Shift Assay of in vitro Phosphorylated RNA Polymerase II Carboxyl-terminal Domain Substrates.
Bio-protocol (2021-03-05)
Joshua E Mayfield, Seema Irani, Yan Zhang
ABSTRACT
Eukaryotic RNA polymerase II transcribes all protein-coding mRNAs and is highly regulated. A key mechanism directing RNA polymerase II and facilitating the co-transcriptional processing of mRNAs is the phosphorylation of its highly repetitive carboxyl-terminal domain (CTD) of its largest subunit, RPB1, at specific residues. A variety of techniques exist to identify and quantify the degree of CTD phosphorylation, including phosphorylation-specific antibodies and mass spectrometry. Electrophoretic mobility shift assays (EMSAs) have been utilized since the discovery of CTD phosphorylation and continue to represent a simple, direct, and widely applicable approach for qualitatively monitoring CTD phosphorylation. We present a standardized method for EMSA analysis of recombinant GST-CTD substrates phosphorylated by a variety of CTD kinases. Strategies to analyze samples under both denatured/reduced and semi-native conditions are provided. This method represents a simple, direct, and reproducible means to monitor CTD phosphorylation in recombinant substrates utilizing equipment common to molecular biology labs and readily applicable to downstream analyses including immunoblotting and mass spectrometry.
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Yeast Extract, for use in microbial growth medium
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%