Merck
  • Development and evaluation of solid self nano-emulsifying formulation of rosuvastatin calcium for improved bioavailability
Development and evaluation of solid self nano-emulsifying formulation of rosuvastatin calcium for improved bioavailability
Tropical Journal of Pharmaceutical Research (2015)
Kulkarni NS, et al.