  • Search Results
  • Assessing kinetics and recruitment of DNA repair factors using high content screens.
Assessing kinetics and recruitment of DNA repair factors using high content screens.
Cell reports (2021-12-30)
Barbara Martinez-Pastor, Giorgia G Silveira, Thomas L Clarke, Dudley Chung, Yuchao Gu, Claudia Cosentino, Lance S Davidow, Gadea Mata, Sylvana Hassanieh, Jayme Salsman, Alberto Ciccia, Narkhyun Bae, Mark T Bedford, Diego Megias, Lee L Rubin, Alejo Efeyan, Graham Dellaire, Raul Mostoslavsky
ABSTRACT
Repair of genetic damage is coordinated in the context of chromatin, so cells dynamically modulate accessibility at DNA breaks for the recruitment of DNA damage response (DDR) factors. The identification of chromatin factors with roles in DDR has mostly relied on loss-of-function screens while lacking robust high-throughput systems to study DNA repair. In this study, we have developed two high-throughput systems that allow the study of DNA repair kinetics and the recruitment of factors to double-strand breaks in a 384-well plate format. Using a customized gain-of-function open-reading frame library ("ChromORFeome" library), we identify chromatin factors with putative roles in the DDR. Among these, we find the PHF20 factor is excluded from DNA breaks, affecting DNA repair by competing with 53BP1 recruitment. Adaptable for genetic perturbations, small-molecule screens, and large-scale analysis of DNA repair, these resources can aid our understanding and manipulation of DNA repair.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-RNF168 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse