Merck
Isolation and Maintenance of Murine Embryonic Striatal Neurons.
Bio-protocol (2018-04-20)
Luana Naia, A Cristina Rego
ABSTRACT
Primary cultures of murine striatal neurons are widely used to explore cellular mechanisms in neurobiology, including brain diseases. Here we describe a detailed and standardized protocol to dissect and culture embryonic murine striatal neurons GABA-positive/DARPP-32-positive for 12 days in vitro, when they show good neuronal cell connectivity and the presence of dendritic spines, which reflects the maturation of the network.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein