Merck
  • Astroglial ER-mitochondria calcium transfer mediates endocannabinoid-dependent synaptic integration.
Astroglial ER-mitochondria calcium transfer mediates endocannabinoid-dependent synaptic integration.
Cell reports (2021-12-23)
Roman Serrat, Ana Covelo, Vladimir Kouskoff, Sebastien Delcasso, Andrea Ruiz-Calvo, Nicolas Chenouard, Carol Stella, Corinne Blancard, Benedicte Salin, Francisca Julio-Kalajzić, Astrid Cannich, Federico Massa, Marjorie Varilh, Severine Deforges, Laurie M Robin, Diego De Stefani, Arnau Busquets-Garcia, Frederic Gambino, Anna Beyeler, Sandrine Pouvreau, Giovanni Marsicano
ABSTRACT
Intracellular calcium signaling underlies the astroglial control of synaptic transmission and plasticity. Mitochondria-endoplasmic reticulum contacts (MERCs) are key determinants of calcium dynamics, but their functional impact on astroglial regulation of brain information processing is unexplored. We found that the activation of astrocyte mitochondrial-associated type-1 cannabinoid (mtCB1) receptors determines MERC-dependent intracellular calcium signaling and synaptic integration. The stimulation of mtCB1 receptors promotes calcium transfer from the endoplasmic reticulum to mitochondria through a specific molecular cascade, involving the mitochondrial calcium uniporter (MCU). Physiologically, mtCB1-dependent mitochondrial calcium uptake determines the dynamics of cytosolic calcium events in astrocytes upon endocannabinoid mobilization. Accordingly, electrophysiological recordings in hippocampal slices showed that conditional genetic exclusion of mtCB1 receptors or dominant-negative MCU expression in astrocytes blocks lateral synaptic potentiation, through which astrocytes integrate the activity of distant synapses. Altogether, these data reveal an endocannabinoid link between astroglial MERCs and the regulation of brain network functions.
Sigma-Aldrich
Monoclonal Anti-S100B antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2720, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse