  • Search Results
  • NETs decorated with bioactive IL-33 infiltrate inflamed tissues and induce IFN-α production in patients with SLE.
NETs decorated with bioactive IL-33 infiltrate inflamed tissues and induce IFN-α production in patients with SLE.
JCI insight (2021-09-24)
Spiros Georgakis, Katerina Gkirtzimanaki, Garyfalia Papadaki, Hariklia Gakiopoulou, Elias Drakos, Maija-Leena Eloranta, Manousos Makridakis, Georgia Kontostathi, Jerome Zoidakis, Eirini Baira, Lars Rönnblom, Dimitrios T Boumpas, Prodromos Sidiropoulos, Panayotis Verginis, George Bertsias
ABSTRACT
IL-33, a nuclear alarmin released during cell death, exerts context-specific effects on adaptive and innate immune cells, eliciting potent inflammatory responses. We screened blood, skin, and kidney tissues from patients with systemic lupus erythematosus (SLE), a systemic autoimmune disease driven by unabated type I IFN production, and found increased amounts of extracellular IL-33 complexed with neutrophil extracellular traps (NETs), correlating with severe, active disease. Using a combination of molecular, imaging, and proteomic approaches, we show that SLE neutrophils, activated by disease immunocomplexes, release IL-33-decorated NETs that stimulate robust IFN-α synthesis by plasmacytoid DCs in a manner dependent on the IL-33 receptor ST2L. IL33-silenced neutrophil-like cells cultured under lupus-inducing conditions generated NETs with diminished interferogenic effect. Importantly, NETs derived from patients with SLE are enriched in mature bioactive isoforms of IL-33 processed by the neutrophil proteases elastase and cathepsin G. Pharmacological inhibition of these proteases neutralized IL-33-dependent IFN-α production elicited by NETs. We believe these data demonstrate a novel role for cleaved IL-33 alarmin decorating NETs in human SLE, linking neutrophil activation, type I IFN production, and end-organ inflammation, with skin pathology mirroring that observed in the kidneys.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Donkey Anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Cathepsin G Inhibitor I - CAS 429676-93-7 - Calbiochem, The Cathepsin G Inhibitor I, also referenced under CAS 429676-93-7, controls the biological activity of Cathepsin G. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sivelestat sodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®