Merck
  • Mouse innate-like B-1 lymphocytes promote inhaled particle-induced in vitro granuloma formation and inflammation in conjunction with macrophages.
Mouse innate-like B-1 lymphocytes promote inhaled particle-induced in vitro granuloma formation and inflammation in conjunction with macrophages.
Archives of toxicology (2021-12-23)
Léa Hiéronimus, Raïssa Demazy, Laura Christiaens, Francine Uwambayinema, Jean-François Geuens, Youssof Yacoub, François Huaux
ABSTRACT
The current paradigm for explaining lung granulomatous diseases induced by inhaled particles is mainly based on macrophages. This mechanism is now challenging because B lymphocytes also infiltrate injured tissue, and the deficiency in B lymphocytes is associated with limited lung granulomas in silica-treated mice. Here, we investigated how B lymphocytes respond to micro- and nanoparticles by combining in vivo and in vitro mouse models. We first demonstrated that innate-like B-1 lymphocytes (not conventional B-2 lymphocytes or plasma cells) specifically accumulated during granuloma formation in mice instilled with crystalline silica (DQ12, 2.5 mg/mouse) and carbon nanotubes (CNT Mitsui, 0.2 mg/mouse). In comparison to macrophages, peritoneal B-1 lymphocytes purified from naïve mice were resistant to the pyroptotic activity of reactive particles (up to 1 mg/mL) but clustered to establish in vitro cell/particle aggregates. Mouse B-1 lymphocytes (not B-2 lymphocytes) in coculture with macrophages and CNT (0.1 µg/mL) organized three-dimensional spheroid structures in Matrigel and stimulated the release of TIMP-1. Furthermore, purified B-1 lymphocytes are sensitive to nanosilica toxicity through radical generation in culture. Nanosilica-exposed B-1 lymphocytes released proinflammatory cytokines and alarmins. In conclusion, our data indicate that in addition to macrophages, B-1 lymphocytes participate in micrometric particle-induced granuloma formation and display inflammatory functions in response to nanoparticles.
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., Molecular Biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein