Merck
  • Ethanol triggers sphingosine 1-phosphate elevation along with neuroapoptosis in the developing mouse brain.
Ethanol triggers sphingosine 1-phosphate elevation along with neuroapoptosis in the developing mouse brain.
Journal of neurochemistry (2012-03-08)
Goutam Chakraborty, Mitsuo Saito, Relish Shah, Rui-Fen Mao, Csaba Vadasz, Mariko Saito
ABSTRACT
Our previous studies have indicated that de novo ceramide synthesis plays a critical role in ethanol-induced apoptotic neurodegeneration in the 7-day-old mouse brain. In this study, we examined whether the formation of sphingosine 1-phosphate (S1P), a ceramide metabolite, is associated with this apoptotic pathway. Analyses of basal levels of S1P-related compounds indicated that S1P, sphingosine, sphingosine kinase 2, and S1P receptor 1 increased significantly during postnatal brain development. In the 7-day-old mouse brain, sphingosine kinase 2 was localized mainly in neurons. Subcellular fractionation studies of the brain homogenates showed that sphingosine kinase 2 was enriched in the plasma membrane and the synaptic membrane/synaptic vesicle fractions, but not in the nuclear and mitochondrial/lysosomal fractions. Ethanol exposure in 7-day-old mice induced sphingosine kinase 2 activation and increased the brain level of S1P transiently 2-4 h after exposure, followed by caspase 3 activation that peaked around 8 h after exposure. Treatment with dimethylsphingosine, an inhibitor of sphingosine kinases, attenuated the ethanol-induced caspase 3 activation and the subsequent neurodegeneration. These results indicate that ethanol activates sphingosine kinase 2, leading to a transient increase in S1P, which may be involved in neuroapoptotic action of ethanol in the developing brain.
Sigma-Aldrich
Sphingosine kinase 2 (long form) Active human, recombinant, expressed in baculovirus, ≥80% (SDS-PAGE)