Merck
  • Binding and dissociation of human topoisomerase I with hairpin-loop RNAs: implications for the regulation of HIV-1 replication
Binding and dissociation of human topoisomerase I with hairpin-loop RNAs: implications for the regulation of HIV-1 replication
Biochemical and Biophysical Research Communications (2012)
Takahashi H, Sawa H, Hasegawa H,