Merck
Multi-CUT&Tag to simultaneously profile multiple chromatin factors.
STAR protocols (2022-02-01)
Sneha Gopalan, Thomas G Fazzio
ABSTRACT
Genome-wide chromatin mapping approaches typically focus on one protein at a time. We recently developed multi-CUT&Tag, which enables simultaneous mapping of multiple chromatin proteins in the same single cells or pools of cells. Using barcoded adapters loaded onto antibody-protein A-Tn5 transposase complexes, multi-CUT&Tag marks the locations of each chromatin protein and directly detects colocalization of different proteins in the same cell(s). Although slightly more laborious than CUT&Tag, multi-CUT&Tag provides a powerful option for generating multi-factor maps for epigenomic profiling. For complete details on the use and execution of this protocol, please refer to Gopalan et al. (2021).
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid tetrasodium salt dihydrate, BioReagent, suitable for cell culture, 98.5-102.0%
SAFC
Spermidine trihydrochloride, ≥99.0% (AT)