Merck
  • A human cornea-on-a-chip for the study of epithelial wound healing by extracellular vesicles.
A human cornea-on-a-chip for the study of epithelial wound healing by extracellular vesicles.
iScience (2022-04-29)
Zitong Yu, Rui Hao, Jing Du, Xiaoliang Wu, Xi Chen, Yi Zhang, Wei Li, Zhongze Gu, Hui Yang
ABSTRACT
Organs-on-chips are microfluidic devices for cell culturing to simulate tissue-level or organ-level physiology and recapitulate their microenvironment, providing new and significant solutions other than traditional animal tests. In vitro testing platforms for ocular biological studies have been increasingly used in preclinical efficacy and toxicity prediction. Here, we developed a microfluidic platform consisting of human corneal cells and porous membrane, replicating the multi-scale structural organization and biological phenotype. We verified the fully integrated human cornea's barrier effects on the chip. Moreover, we found that extracellular vesicles derived from bone marrow-derived mesenchymal stem cells can significantly accelerate the mild corneal epithelial wound healing, and the decreased expression of matrix metallopeptidase-2 protein indicated that this method effectively inhibits corneal inflammation and angiogenesis. This work improves our ability to simulate the interaction between the human cornea and the external world in vitro and contributes to the future development of new screening platforms for biopharmaceuticals.
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Transferrin human, powder, BioReagent, suitable for cell culture