Merck
  • Melanoma secretion of transforming growth factor-β2 leads to loss of epidermal AMBRA1 threatening epidermal integrity and facilitating tumour ulceration.
Melanoma secretion of transforming growth factor-β2 leads to loss of epidermal AMBRA1 threatening epidermal integrity and facilitating tumour ulceration.
The British journal of dermatology (2021-11-14)
I Cosgarea, A T McConnell, T Ewen, D Tang, D S Hill, M Anagnostou, M Elias, R A Ellis, A Murray, L C Spender, P Giglio, M Gagliardi, A Greenwood, M Piacentini, G J Inman, G M Fimia, M Corazzari, J L Armstrong, P E Lovat
ABSTRACT
For patients with early American Joint Committee on Cancer (AJCC)-stage melanoma the combined loss of the autophagy regulatory protein AMBRA1 and the terminal differentiation marker loricrin in the peritumoral epidermis is associated with a significantly increased risk of metastasis. The aim of the present study was to evaluate the potential contribution of melanoma paracrine transforming growth factor (TGF)-β signalling to the loss of AMBRA1 in the epidermis overlying the primary tumour and disruption of epidermal integrity. Immunohistochemistry was used to analyse AMBRA1 and TGF-β2 in a cohort of 109 AJCC all-stage melanomas, and TGF-β2 and claudin-1 in a cohort of 30 or 42 AJCC stage I melanomas, respectively, with known AMBRA1 and loricrin (AMLo) expression. Evidence of pre-ulceration was analysed in a cohort of 42 melanomas, with TGF-β2 signalling evaluated in primary keratinocytes. Increased tumoral TGF-β2 was significantly associated with loss of peritumoral AMBRA1 (P < 0·05), ulceration (P < 0·001), AMLo high-risk status (P < 0·05) and metastasis (P < 0·01). TGF-β2 treatment of keratinocytes resulted in downregulation of AMBRA1, loricrin and claudin-1, while knockdown of AMBRA1 was associated with decreased expression of claudin-1 and increased proliferation of keratinocytes (P < 0·05). Importantly, we show loss of AMBRA1 in the peritumoral epidermis was associated with decreased claudin-1 expression (P < 0·05), parakeratosis (P < 0·01) and cleft formation in the dermoepidermal junction (P < 0·05). Collectively, these data suggest a paracrine mechanism whereby TGF-β2 causes loss of AMBRA1 overlying high-risk AJCC early-stage melanomas and reduced epidermal integrity, thereby facilitating erosion of the epidermis and tumour ulceration.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Transforming Growth Factor-β2 human, TGF-β2, recombinant, expressed in NSO cells, powder, suitable for cell culture
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture