Merck
  • In vivo topology converts competition for cell-matrix adhesion into directional migration.
In vivo topology converts competition for cell-matrix adhesion into directional migration.
Nature communications (2019-04-05)
Fernanda Bajanca, Nadège Gouignard, Charlotte Colle, Maddy Parsons, Roberto Mayor, Eric Theveneau
ABSTRACT
When migrating in vivo, cells are exposed to numerous conflicting signals: chemokines, repellents, extracellular matrix, growth factors. The roles of several of these molecules have been studied individually in vitro or in vivo, but we have yet to understand how cells integrate them. To start addressing this question, we used the cephalic neural crest as a model system and looked at the roles of its best examples of positive and negative signals: stromal-cell derived factor 1 (Sdf1/Cxcl12) and class3-Semaphorins. Here we show that Sdf1 and Sema3A antagonistically control cell-matrix adhesion via opposite effects on Rac1 activity at the single cell level. Directional migration at the population level emerges as a result of global Semaphorin-dependent confinement and broad activation of adhesion by Sdf1 in the context of a biased Fibronectin distribution. These results indicate that uneven in vivo topology renders the need for precise distribution of secreted signals mostly dispensable.
Product Description
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
O9-1 Mouse Cranial Neural Crest Cell Line, stably expresses stem cell markers and neural crest markers