Merck
  • Human induced-T-to-natural killer cells have potent anti-tumour activities.
Human induced-T-to-natural killer cells have potent anti-tumour activities.
Biomarker research (2022-03-26)
Zhiwu Jiang, Le Qin, Yuou Tang, Rui Liao, Jingxuan Shi, Bingjia He, Shanglin Li, Diwei Zheng, Yuanbin Cui, Qiting Wu, Youguo Long, Yao Yao, Zhihui Wei, Qilan Hong, Yi Wu, Yuanbang Mai, Shixue Gou, Xiaoping Li, Robert Weinkove, Sam Norton, Wei Luo, Weineng Feng, Hongsheng Zhou, Qifa Liu, Jiekai Chen, Liangxue Lai, Xinwen Chen, Duanqing Pei, Thomas Graf, Xingguo Liu, Yangqiu Li, Pentao Liu, Zhenfeng Zhang, Peng Li
ABSTRACT
Adoptive cell therapy (ACT) is a particularly promising area of cancer immunotherapy, engineered T and NK cells that express chimeric antigen receptors (CAR) are being explored for treating hematopoietic malignancies but exhibit limited clinical benefits for solid tumour patients, successful cellular immunotherapy of solid tumors demands new strategies. Inactivation of BCL11B were performed by CRISPR/Cas9 in human T cells. Immunophenotypic and transcriptional profiles of sgBCL11B T cells were characterized by cytometer and transcriptomics, respectively. sgBCL11B T cells are further engineered with chimeric antigen receptor. Anti-tumor activity of ITNK or CAR-ITNK cells were evaluated in preclinical and clinical studies. We report that inactivation of BCL11B in human CD8+ and CD4+ T cells induced their reprogramming into induced T-to-natural killer cells (ITNKs). ITNKs contained a diverse TCR repertoire; downregulated T cell-associated genes such as TCF7 and LEF1; and expressed high levels of NK cell lineage-associated genes. ITNKs and chimeric antigen receptor (CAR)-transduced ITNKs selectively lysed a variety of cancer cells in culture and suppressed the growth of solid tumors in xenograft models. In a preliminary clinical study, autologous administration of ITNKs in patients with advanced solid tumors was well tolerated, and tumor stabilization was seen in six out nine patients, with one partial remission. The novel ITNKs thus may be a promising novel cell source for cancer immunotherapy. ClinicalTrials.gov, NCT03882840 . Registered 20 March 2019-Retrospectively registered.
Product Description
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel, Premixed 96-well Human High Sensitivity T Cell 21-Plex Panel