Merck
  • Promoting antigen escape from dendritic cell endosomes potentiates anti-tumoral immunity.
Promoting antigen escape from dendritic cell endosomes potentiates anti-tumoral immunity.
Cell reports. Medicine (2022-05-03)
Jean-Pierre Bikorimana, Natasha Salame, Simon Beaudoin, Mohammad Balood, Théo Crosson, Jamilah Abusarah, Sebastien Talbot, Raimar Löbenberg, Sebastien Plouffe, Moutih Rafei
ABSTRACT
The cross-presenting capacity of dendritic cells (DCs) can be limited by non-specific degradation during endosome maturation. To bypass this limitation, we present in this study a new Accum-based formulation designed to promote endosome-to-cytosol escape. Treatment of primary DCs with Accum linked to the xenoantigen ovalbumin (OVA) triggers endosomal damages and enhances protein processing. Despite multiple challenges using ascending doses of tumor cells, DC prophylactic vaccination results in complete protection due to increased levels of effector CD4 and CD8 T cells as well as high production of pro-inflammatory mediators. When combined with anti-PD-1, therapeutic vaccination using both syngeneic and allogeneic Accum-OVA-pulsed DCs triggers potent anti-tumoral responses. The net outcome culminates in increased CD11c, CD8, and NK infiltration along with a high CD8/Treg ratio. These highly favorable therapeutic effects highlight the promising potential of Accum as a distinct and potent technology platform suitable for the design of next generation cell cancer vaccines.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Collagenase Type IV, Cls IV
Roche
Collagenase D, from Clostridium histolyticum