Merck
  • Snail Mucus Protective Effect on Ethanol-Induced Gastric Ulcers in Mice.
Snail Mucus Protective Effect on Ethanol-Induced Gastric Ulcers in Mice.
Life (Basel, Switzerland) (2022-07-28)
Lubomir Petrov, Mihail Kachaunov, Albena Alexandrova, Elina Tsvetanova, Almira Georgieva, Aleksander Dolashki, Lyudmila Velkova, Pavlina Dolashka
ABSTRACT
Nowadays, an increased interest in natural compounds with preventive or therapeutic potential for various diseases has been observed. Given the involvement of oxidative stress in the pathogenesis of gastric ulcer (GU) and the wide range of bioactive compounds isolated from snails, this study aimed to investigate the protective effect of Cornu aspersum (Müller, 1774) mucus on ethanol-induced GUs. Male albino mice were divided into Control, Ethanol, Mucus + Ethanol and Mucus + Omeprazole treated groups. The GUs were induced by administration of 96% ethanol (10 mL/kg, per os). One hour before ulcer induction, the mice of Mucus + Ethanol group were pretreated with mucus (20 mg/kg, per os), and the mice of Mucus + Omeprazole group were pretreated with omeprazole (20 mg/kg, per os). Ethanol administration caused grave lesions of gastric mucosa and a significant decrease of glutathione (GSH) and superoxide dismutase (SOD), catalase, and glutathione reductase (GR) activities. In the animals with mucus or omeprazole pre-administration compared to the Ethanol group, the following were observed: only a small number of hemorrhagic fields, significantly reduced GU index with calculated 73% protection by mucus and 78% protection by omeprazole, and significant recovery of mucosal GSH and SOD and GR activities. In addition, the mucus inhibited Helicobacter pylori growth. Thus, the protective effect of C. aspersum mucus on both gastric mucosa and gastric antioxidant potential in ethanol-induced GU model suggests that it may serve as a good tool for prevention of this disease.
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
Sigma-Aldrich
Lipid Peroxidation (MDA) Assay Kit, sufficient for 100 colorimetric or fluorometric tests
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells