Merck
  • A pro-tumorigenic mDia2-MIRO1 axis controls mitochondrial positioning and function in cancer-associated fibroblasts.
A pro-tumorigenic mDia2-MIRO1 axis controls mitochondrial positioning and function in cancer-associated fibroblasts.
Cancer research (2022-08-24)
Michael Cangkrama, Huan Liu, James Whipman, Maria Zubair, Mai Matsushita, Michela Di Filippo, Manfred Kopf, Metello Innocenti, Sabine Werner
ABSTRACT
Cancer-associated fibroblasts (CAF) are key regulators of tumorigenesis. Further insights into the tumor promoting mechanisms of action of CAFs could help improve cancer diagnosis and treatment. Here we show that the formin mDia2 regulates the positioning and function of mitochondria in dermal fibroblasts, thereby promoting a pro-tumorigenic CAF phenotype. Mechanistically, mDia2 stabilized the mitochondrial trafficking protein MIRO1. Loss of mDia2 or MIRO1 in fibroblasts or CAFs reduced the presence of mitochondria and ATP levels near the plasma membrane and at CAF-tumor cell contact sites, caused metabolic alterations characteristic of mitochondrial dysfunction, and suppressed the secretion of pro-tumorigenic proteins. In mouse models of squamous carcinogenesis, genetic or pharmacological inhibition of mDia2, MIRO1, or their common upstream regulator activin A inhibited tumor formation. Consistently, co-upregulation of mDia2 and MIRO1 in the stroma of various human cancers negatively correlated with survival. This work unveils a key role of mitochondria in the pro-tumorigenic CAF phenotype and identifies an activin A/mDia2/MIRO1 signaling axis in CAFs with diagnostic and therapeutic potential.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
SMIFH2, ≥98% (HPLC)
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit