Merck
  • Balance between immunoregulatory B cells and plasma cells drives pancreatic tumor immunity.
Balance between immunoregulatory B cells and plasma cells drives pancreatic tumor immunity.
Cell reports. Medicine (2022-09-14)
Bhalchandra Mirlekar, Yan Wang, Sirui Li, Mi Zhou, Sarah Entwistle, Tristan De Buysscher, Ashley Morrison, Gabriela Herrera, Cameron Harris, Benjamin G Vincent, Jenny P-Y Ting, Naim Rashid, William Y Kim, Jen Jen Yeh, Yuliya Pylayeva-Gupta
ABSTRACT
Plasma cell responses are associated with anti-tumor immunity and favorable response to immunotherapy. B cells can amplify anti-tumor immune responses through antibody production; yet B cells in patients and tumor-bearing mice often fail to support this effector function. We identify dysregulated transcriptional program in B cells that disrupts differentiation of naive B cells into anti-tumor plasma cells. The signaling network contributing to this dysfunction is driven by interleukin (IL) 35 stimulation of a STAT3-PAX5 complex that upregulates the transcriptional regulator BCL6 in naive B cells. Transient inhibition of BCL6 in tumor-educated naive B cells is sufficient to reverse the dysfunction in B cell differentiation, stimulating the intra-tumoral accumulation of plasma cells and effector T cells and rendering pancreatic tumors sensitive to anti-programmed cell death protein 1 (PD-1) blockade. Our findings argue that B cell effector dysfunction in cancer can be due to an active systemic suppression program that can be targeted to synergize with T cell-directed immunotherapy.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Anti-EBI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution