Merck
Improved Sendai viral system for reprogramming to naive pluripotency.
Cell reports methods (2022-12-01)
Akira Kunitomi, Ryoko Hirohata, Vanessa Arreola, Mitsujiro Osawa, Tomoaki M Kato, Masaki Nomura, Jitsutaro Kawaguchi, Hiroto Hara, Kohji Kusano, Yasuhiro Takashima, Kazutoshi Takahashi, Keiichi Fukuda, Naoko Takasu, Shinya Yamanaka
ABSTRACT
Naive human induced pluripotent stem cells (iPSCs) can be generated by reprogramming somatic cells with Sendai virus (SeV) vectors. However, only dermal fibroblasts have been successfully reprogrammed this way, and the process requires culture on feeder cells. Moreover, SeV vectors are highly persistent and inhibit subsequent differentiation of iPSCs. Here, we report a modified SeV vector system to generate transgene-free naive human iPSCs with superior differentiation potential. The modified method can be applied not only to fibroblasts but also to other somatic cell types. SeV vectors disappear quickly at early passages, and this approach enables the generation of naive iPSCs in a feeder-free culture. The naive iPSCs generated by this method show better differentiation to trilineage and extra-embryonic trophectoderm than those derived by conventional methods. This method can expand the application of iPSCs to research on early human development and regenerative medicine.
Sigma-Aldrich
Gö 6983, ≥97%
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).