Merck
  • PepDistiller: A quality control tool to improve the sensitivity and accuracy of peptide identifications in shotgun proteomics.
PepDistiller: A quality control tool to improve the sensitivity and accuracy of peptide identifications in shotgun proteomics.
Proteomics (2012-05-25)
Ning Li, Songfeng Wu, Chengpu Zhang, Cheng Chang, Jiyang Zhang, Jie Ma, Liwei Li, Xiaohong Qian, Ping Xu, Yunping Zhu, Fuchu He
ABSTRACT
In this study, we presented a quality control tool named PepDistiller to facilitate the validation of MASCOT search results. By including the number of tryptic termini, and integrating a refined false discovery rate (FDR) calculation method, we demonstrated the improved sensitivity of peptide identifications obtained from semitryptic search results. Based on the analysis of a complex data set, approximately 7% more peptide identifications were obtained using PepDistiller than using MASCOT Percolator. Moreover, the refined method generated lower FDR estimations than the percentage of incorrect target (PIT) fixed method applied in Percolator. Using a standard data set, we further demonstrated the increased accuracy of the refined FDR estimations relative to the PIT-fixed FDR estimations. PepDistiller is fast and convenient to use, and is freely available for academic access. The software can be downloaded from http://www.bprc.ac.cn/pepdistiller.
Sigma-Aldrich
Universal Proteomics Standard Set, Protein Mass Spectrometry Calibration Standard