Merck
  • Electrophysiological characterization of voltage-gated Na+ current expressed in the highly metastatic Mat-LyLu cell line of rat prostate cancer.
Electrophysiological characterization of voltage-gated Na+ current expressed in the highly metastatic Mat-LyLu cell line of rat prostate cancer.
Journal of cellular physiology (1998-03-10)
J A Grimes, M B Djamgoz
ABSTRACT
Voltage-gated Na+ channels, classically associated with impulse conduction in excitable tissues, are also found in a variety of epithelial cell types where their possible functions are not known so well. We have previously reported expression of a voltage-gated Na+ channel specifically in the highly metastatic Mat-LyLu rat prostate cancer cell line; blockage of the current with tetrodotoxin (TTX) significantly reduced the invasiveness of the cells in vitro, suggesting that the channel may have a functional role in metastasis. The aim of the present study was to characterize this current using the whole-cell patch clamp recording technique, and compare it to Na+ currents found in various other tissues. The inward current of the Mat-LyLu cells was abolished completely, but reversibly, in Na+-free solution, confirming that Na+ was indeed the permeant ion. Activation occurred at -40 mV and currents reached a maximal amplitude at around 6 mV. Boltzmann fits to current activation and steady-state inactivation revealed that the currents were half activated at about -15 mV and half inactivated at -80 mV. Both current inactivation and recovery from inactivation followed a double-exponential time course with fast and slow components. The Na+ currents were highly sensitive to block by TTX (IC50 approximately 18 nM), whilst 1 microM mu-conotoxin GIIIA mostly had no effect. 100 microM Cd2+ also had no effect on the current, whilst 2.5 mM Cd2+, Mn2+, and Co2+ each caused a depolarizing shift in activation and a reduction in peak conductance of around 20%. In conclusion, the Na+ channel expressed in the highly metastatic Mat-LyLu cell line appeared to have electrophysiological and pharmacological properties of TTX-sensitive channels. Further work is needed, however, to elucidate the exact nature of the channel protein and the mechanism(s) of its involvement in cellular invasiveness.