Merck
  • Gingerenone A Induces Antiproliferation and Senescence of Breast Cancer Cells.
Gingerenone A Induces Antiproliferation and Senescence of Breast Cancer Cells.
Antioxidants (Basel, Switzerland) (2022-03-26)
Tzu-Jung Yu, Jen-Yang Tang, Jun-Ping Shiau, Ming-Feng Hou, Chia-Hung Yen, Fu Ou-Yang, Chung-Yi Chen, Hsueh-Wei Chang
ABSTRACT
Ginger is a popular spice and consists of several bioactive antioxidant compounds. Gingerenone A (Gin A), a novel compound isolated from Zingiber officinale, is rarely investigated for its anti-breast-cancer properties. Some ginger extracts have been reported to initiate senescence, an anticancer strategy. However, the anticancer effects of Gin A on breast cancer cells remain unclear. The present study aims to assess the modulating impact of Gin A acting on proliferation and senescence to breast cancer cells. Gin A diminished the cellular ATP content and decreased the cell viability of the MTS assay in several breast cancer cell lines. It also showed a delayed G2/M response to breast cancer cells (MCF7 and MDA-MB-231). N-acetylcysteine (NAC), an oxidative stress inhibitor, can revert these responses of antiproliferation and G2/M delay. The oxidative stress and senescence responses of Gin A were further validated by increasing reactive oxygen species, mitochondrial superoxide, and β-galactosidase activity, which were reverted by NAC. Gin A also upregulated senescence-associated gene expressions. In addition to oxidative stress, Gin A also induced DNA damage responses by increasing γH2AX level and foci and generating 8-hydroxyl-2'-deoxyguanosine in breast cancer cells, which were reverted by NAC. Therefore, Gin A promotes antiproliferation and senescence of breast cancer cells induced by oxidative stress.
Sigma-Aldrich
Gingerenone A, ≥95% (HPLC)