Merck
  • Inhibiting neuronal AC1 for treating anxiety and headache in the animal model of migraine.
Inhibiting neuronal AC1 for treating anxiety and headache in the animal model of migraine.
iScience (2023-05-26)
Ren-Hao Liu, Mingjie Zhang, Man Xue, Tao Wang, Jing-Shan Lu, Xu-Hui Li, Yu-Xin Chen, Kexin Fan, Wantong Shi, Si-Bo Zhou, Qi-Yu Chen, Li Kang, Qian Song, Shengyuan Yu, Min Zhuo
ABSTRACT
Migraines are a common medical condition. From a basic science point of view, the central mechanism for migraine and headache is largely unknown. In the present study, we demonstrate that cortical excitatory transmission is significantly enhanced in the anterior cingulate cortex (ACC)-a brain region which is critical for pain perception. Biochemical studies found that the phosphorylation levels of both the NMDA receptor GluN2B and AMPA receptor GluA1 were enhanced in ACC of migraine rats. Both the presynaptic release of glutamate and postsynaptic responses of AMPA receptors and NMDA receptors were enhanced. Synaptic long-term potentiation (LTP) was occluded. Furthermore, behavioral anxiety and nociceptive responses were increased, which were reversed by application of AC1 inhibitor NB001 within ACC. Our results provide strong evidence that cortical LTPs contribute to migraine-related pain and anxiety. Drugs that inhibit cortical excitation such as NB001 may serve as potential medicines for treating migraine in the future.
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NMDAR2A Antibody, serum, Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Bradykinin acetate salt, powder, ≥98% (HPLC)
Sigma-Aldrich
Anti-NR1 Antibody, CT, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-NR2B (Ser1303) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 831, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Glutamate Receptor 1 Antibody, phosphoSer 845, Chemicon®, from rabbit
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-NMDAR2B Antibody, pain, serum, Chemicon®