Merck
  • LSD1 interacting with HSP90 promotes skin wound healing by inducing metabolic reprogramming of hair follicle stem cells through the c-MYC/LDHA axis.
LSD1 interacting with HSP90 promotes skin wound healing by inducing metabolic reprogramming of hair follicle stem cells through the c-MYC/LDHA axis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2023-06-21)
Shuiqi Li, Jie Yu, Jiangan Zhang, Xiaohong Li, Jianbin Yu
ABSTRACT
It has been demonstrated that hair follicle stem cells (HFSCs) can contribute to wound closure and repair. However, the specific mechanism remains unclear due to the complexity of the wound repair process. Lysine-specific demethylase 1 (LSD1), an important gene for the regulation of stem cell differentiation, has been reported to participate in wound healing regulation. Heat shock protein 90 (HSP90), a chaperone protein, is recently discovered to be a driver gene for wound healing. This study explored the molecular mechanisms by which the binding between LSD1 and HSP90 affects the role of HFSCs during skin wound healing. Following bioinformatics analysis, the key genes acting on HFSCs were identified. The expression of LSD1, HSP90, and c-MYC was found to be upregulated in differentiated HFSCs. Analysis of their binding affinity revealed that LSD1 interacted with HSP90 to enhance the stability of the transcription factor c-MYC. Lactate dehydrogenase A (LDHA) has been documented to be essential for HFSC activation. Therefore, we speculate that LDHA may induce the differentiation of HFSCs through glucose metabolism reprogramming. The results showed that c-MYC activated LDHA activity to promote glycolytic metabolism, proliferation, and differentiation of HFSCs. Finally, in vivo animal experiments further confirmed that LSD1 induced skin wound healing in mice via the HSP90/c-MYC/LDHA axis. From our data, we conclude that LSD1 interacting with HSP90 accelerates skin wound healing by inducing HFSC glycolytic metabolism, proliferation, and differentiation via c-MYC/LDHA axis.
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
Transferrin human, powder, BioReagent, suitable for cell culture