Merck
  • Intercellular transfer of miR-200c-3p impairs the angiogenic capacity of cardiac endothelial cells.
Intercellular transfer of miR-200c-3p impairs the angiogenic capacity of cardiac endothelial cells.
Molecular therapy : the journal of the American Society of Gene Therapy (2022-03-13)
Lara Ottaviani, Rio P Juni, Ricardo C de Abreu, Marida Sansonetti, Vasco Sampaio-Pinto, Julie Halkein, Jana C Hegenbarth, Nadja Ring, Kevin Knoops, Jordy M M Kocken, Carlos de Jesus, Auriane C Ernault, Hamid El Azzouzi, Frank Rühle, Servé Olieslagers, Hugo Fernandes, Lino Ferreira, Luca Braga, Monika Stoll, Diana S Nascimento, Leon J de Windt, Paula A da Costa Martins
ABSTRACT
As mediators of intercellular communication, extracellular vesicles containing molecular cargo, such as microRNAs, are secreted by cells and taken up by recipient cells to influence their cellular phenotype and function. Here we report that cardiac stress-induced differential microRNA content, with miR-200c-3p being one of the most enriched, in cardiomyocyte-derived extracellular vesicles mediates functional cross-talk with endothelial cells. Silencing of miR-200c-3p in mice subjected to chronic increased cardiac pressure overload resulted in attenuated hypertrophy, smaller fibrotic areas, higher capillary density, and preserved cardiac ejection fraction. We were able to maximally rescue microvascular and cardiac function with very low doses of antagomir, which specifically silences miR-200c-3p expression in non-myocyte cells. Our results reveal vesicle transfer of miR-200c-3p from cardiomyocytes to cardiac endothelial cells, underlining the importance of cardiac intercellular communication in the pathophysiology of heart failure.
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder