Merck
  • High-throughput, targeted MHC class I immunopeptidomics using a functional genetics screening platform.
High-throughput, targeted MHC class I immunopeptidomics using a functional genetics screening platform.
Nature biotechnology (2023-01-03)
Peter M Bruno, Richard T Timms, Nouran S Abdelfattah, Yumei Leng, Felipe J N Lelis, Duane R Wesemann, Xu G Yu, Stephen J Elledge
ABSTRACT
Identification of CD8+ T cell epitopes is critical for the development of immunotherapeutics. Existing methods for major histocompatibility complex class I (MHC class I) ligand discovery are time intensive, specialized and unable to interrogate specific proteins on a large scale. Here, we present EpiScan, which uses surface MHC class I levels as a readout for whether a genetically encoded peptide is an MHC class I ligand. Predetermined starting pools composed of >100,000 peptides can be designed using oligonucleotide synthesis, permitting large-scale MHC class I screening. We exploit this programmability of EpiScan to uncover an unappreciated role for cysteine that increases the number of predicted ligands by 9-21%, reveal affinity hierarchies by analysis of biased anchor peptide libraries and screen viral proteomes for MHC class I ligands. Using these data, we generate and iteratively refine peptide binding predictions to create EpiScan Predictor. EpiScan Predictor performs comparably to other state-of-the-art MHC class I peptide binding prediction algorithms without suffering from underrepresentation of cysteine-containing peptides. Thus, targeted immunopeptidomics using EpiScan will accelerate CD8+ T cell epitope discovery toward the goal of individual-specific immunotherapeutics.
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-ERAP1 Antibody, clone 16A7.1, clone 16A7.1, from mouse